Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTGGTGTATTCTTGCAACTGA[C/T]AAAAGGTCAGGGTGGTTGGATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM149B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM149B1 - family with sequence similarity 149 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173348.1 | Intron | NP_775483.1 | ||||
XM_005269744.2 | Intron | XP_005269801.1 | ||||
XM_005269745.2 | Intron | XP_005269802.1 | ||||
XM_005269747.3 | Intron | XP_005269804.1 | ||||
XM_011539737.2 | Intron | XP_011538039.1 | ||||
XM_011539740.2 | Intron | XP_011538042.1 | ||||
XM_017016164.1 | Intron | XP_016871653.1 | ||||
XM_017016165.1 | Intron | XP_016871654.1 | ||||
XM_017016166.1 | Intron | XP_016871655.1 | ||||
XM_017016167.1 | Intron | XP_016871656.1 |