Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCTCCAGTGATGTACATCACA[A/G]CCATGAGGAAGAACCAGCCCATCTG
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 607945 | ||||||||||||||||||||||||||
Literature Links: |
ADIPOR1 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ADIPOR1 - adiponectin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290553.1 | 1226 | Intron | NP_001277482.1 | |||
NM_001290557.1 | 1226 | Missense Mutation | GCT,GTT | A,V 307 | NP_001277486.1 | |
NM_001290629.1 | 1226 | Intron | NP_001277558.1 | |||
NM_015999.5 | 1226 | Missense Mutation | GCT,GTT | A,V 307 | NP_057083.2 | |
XM_017001407.1 | 1226 | Intron | XP_016856896.1 |
Set Membership: |
HapMap |