Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAAGAGATAACATCTATAAACT[A/T]CTCAACTATTAGCATGTACTCAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613293 | ||||||||||||||||||||
Literature Links: |
SH3PXD2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH3PXD2B - SH3 and PX domains 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017995.2 | Intron | NP_001017995.1 | ||||
NM_001308175.1 | Intron | NP_001295104.1 | ||||
XM_017009351.1 | Intron | XP_016864840.1 |