Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTAGGGTCTGACTTCATGGCCA[C/T]GTGAATGCTGACCACAGGGAGCATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 312040 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
POLA1 - DNA polymerase alpha 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016937.3 | Intron | NP_058633.2 | ||||
XM_005274552.3 | Intron | XP_005274609.1 | ||||
XM_006724499.2 | Intron | XP_006724562.1 | ||||
XM_011545540.2 | Intron | XP_011543842.1 | ||||
XM_011545541.2 | Intron | XP_011543843.1 | ||||
XM_017029594.1 | Intron | XP_016885083.1 | ||||
XM_017029595.1 | Intron | XP_016885084.1 | ||||
XM_017029596.1 | Intron | XP_016885085.1 |
Set Membership: |
HapMap |