Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGATGGCTTCTTCTTAACCTC[C/T]TCGGGTGAGTCGAAGGTTTACCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601879 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LGALS9 - galectin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002308.3 | Intron | NP_002299.2 | ||||
NM_009587.2 | Intron | NP_033665.1 | ||||
XM_006721892.2 | Intron | XP_006721955.1 | ||||
XM_006721893.2 | Intron | XP_006721956.1 | ||||
XM_006721894.3 | Intron | XP_006721957.1 | ||||
XM_006721895.3 | Intron | XP_006721958.1 | ||||
XM_011524796.1 | Intron | XP_011523098.1 | ||||
XM_017024623.1 | Intron | XP_016880112.1 |
Set Membership: |
HapMap |