Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAACCTGTGAAAATGCAGATAC[C/T]TGGGCCCTATACCTAGAGATTCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601874 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELL2 - elongation factor for RNA polymerase II 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012081.5 | 4604 | UTR 3 | NP_036213.2 | |||
XM_006714575.2 | 4604 | UTR 3 | XP_006714638.1 | |||
XM_017009239.1 | 4604 | Intron | XP_016864728.1 | |||
XM_017009240.1 | 4604 | UTR 3 | XP_016864729.1 | |||
XM_017009241.1 | 4604 | UTR 3 | XP_016864730.1 | |||
XM_017009242.1 | 4604 | UTR 3 | XP_016864731.1 | |||
XM_017009243.1 | 4604 | UTR 3 | XP_016864732.1 |
Set Membership: |
HapMap |