Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACAGCACTATTCTGGATGCTAG[C/T]GTTTCTCTAACAGCCTAGCAAAAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615564 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM45A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM45A - family with sequence similarity 45 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFXN4 - sideroflexin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213649.1 | 920 | Silent Mutation | ACA,ACG | T,T 249 | NP_998814.1 | |
XM_005269525.4 | 920 | Silent Mutation | ACA,ACG | T,T 240 | XP_005269582.1 | |
XM_005269526.2 | 920 | Silent Mutation | ACA,ACG | T,T 133 | XP_005269583.1 | |
XM_005269527.1 | 920 | Silent Mutation | ACA,ACG | T,T 133 | XP_005269584.1 | |
XM_011539282.2 | 920 | Silent Mutation | ACA,ACG | T,T 133 | XP_011537584.1 |
Set Membership: |
HapMap |