Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGACCTAGAGACATGAGGGCATG[C/G]AGTGAGATGAGCACCCTCCACTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138130 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLUD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLUD1 - glutamate dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318900.1 | Intron | NP_001305829.1 | ||||
NM_001318901.1 | Intron | NP_001305830.1 | ||||
NM_001318902.1 | Intron | NP_001305831.1 | ||||
NM_001318904.1 | Intron | NP_001305833.1 | ||||
NM_001318905.1 | Intron | NP_001305834.1 | ||||
NM_001318906.1 | Intron | NP_001305835.1 | ||||
NM_005271.4 | Intron | NP_005262.1 |
Set Membership: |
HapMap |