Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGGAGAGAAAGCCTCGAGGAA[A/G]CCGTGGAGACGAAATTGAGCAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602054 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBX1 - T-box 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005992.1 | Intron | NP_005983.1 | ||||
NM_080646.1 | Intron | NP_542377.1 | ||||
NM_080647.1 | Intron | NP_542378.1 | ||||
XM_006724312.2 | Intron | XP_006724375.1 | ||||
XM_017028925.1 | Intron | XP_016884414.1 | ||||
XM_017028926.1 | Intron | XP_016884415.1 | ||||
XM_017028927.1 | Intron | XP_016884416.1 | ||||
XM_017028928.1 | Intron | XP_016884417.1 |