Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
QRSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
QRSL1 - glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018292.4 | Intron | NP_060762.3 | ||||
XM_011535924.2 | Intron | XP_011534226.1 |
RTN4IP1 - reticulon 4 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318746.1 | Intron | NP_001305675.1 | ||||
NM_032730.5 | Intron | NP_116119.2 | ||||
XM_011536192.2 | Intron | XP_011534494.1 | ||||
XM_017011375.1 | Intron | XP_016866864.1 | ||||
XM_017011376.1 | Intron | XP_016866865.1 |