Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGGTGAAACTGATCTTGGAGAG[C/T]TGGGAGATTTGCCTTAGCACATCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609389 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5F PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INPP5F - inositol polyphosphate-5-phosphatase F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243194.1 | Intron | NP_001230123.1 | ||||
NM_001243195.1 | Intron | NP_001230124.1 | ||||
NM_014937.3 | Intron | NP_055752.1 | ||||
XM_006717720.3 | Intron | XP_006717783.1 | ||||
XM_011539524.2 | Intron | XP_011537826.1 | ||||
XM_011539525.2 | Intron | XP_011537827.1 | ||||
XM_011539527.2 | Intron | XP_011537829.2 | ||||
XM_011539528.2 | Intron | XP_011537830.1 | ||||
XM_011539529.1 | Intron | XP_011537831.1 | ||||
XM_017015935.1 | Intron | XP_016871424.1 | ||||
XM_017015936.1 | Intron | XP_016871425.1 |
Set Membership: |
HapMap |