Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCTCTGGATTCAGGGAAACCTA[A/G]GGAAGCCAGGCAAGAAGGGTCCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604223 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARPC1B - actin related protein 2/3 complex subunit 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005720.3 | Intron | NP_005711.1 | ||||
XM_006715825.1 | Intron | XP_006715888.1 | ||||
XM_006715826.1 | Intron | XP_006715889.1 |
Set Membership: |
HapMap |