Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTTGCTGAATCTGGTTCCAC[A/G]TTGGTAGATTTTTAAAAAGTTAATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124010 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP3A4 - cytochrome P450 family 3 subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202855.2 | 2235 | Intron | NP_001189784.1 | |||
NM_017460.5 | 2235 | Intron | NP_059488.2 | |||
XM_017011780.1 | 2235 | UTR 3 | XP_016867269.1 |
Set Membership: |
HapMap |