Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGGACGGAATTCGGGACCGACT[A/G]ACGGCCGGCCGGCTTCCCGGAACTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608642 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTGR2 - prostaglandin reductase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF410 - zinc finger protein 410 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242924.1 | 272 | UTR 5 | NP_001229853.1 | |||
NM_001242926.1 | 272 | UTR 5 | NP_001229855.1 | |||
NM_001242927.1 | 272 | UTR 5 | NP_001229856.1 | |||
NM_001242928.1 | 272 | UTR 5 | NP_001229857.1 | |||
NM_021188.2 | 272 | UTR 5 | NP_067011.1 |
Set Membership: |
HapMap |