Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACAGCCTTCTTCAGCATAATAT[A/T]GTCTCCAAAAAAGGCATTTTTTAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607204 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PUM1 - pumilio RNA binding family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001020658.1 | 4334 | UTR 3 | NP_001018494.1 | |||
NM_014676.2 | 4334 | UTR 3 | NP_055491.1 |
SNORD103A - small nucleolar RNA, C/D box 103A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |