Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCACTTTGCCTACAACAATGC[A/G]GAGGAATAAAAAACATGCTTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDBF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZDBF2 - zinc finger DBF-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285549.1 | Intron | NP_001272478.1 | ||||
NM_020923.2 | Intron | NP_065974.1 | ||||
XM_005246711.3 | Intron | XP_005246768.1 | ||||
XM_005246712.4 | Intron | XP_005246769.1 | ||||
XM_005246713.3 | Intron | XP_005246770.1 | ||||
XM_006712656.3 | Intron | XP_006712719.1 | ||||
XM_011511532.2 | Intron | XP_011509834.1 | ||||
XM_011511533.2 | Intron | XP_011509835.1 | ||||
XM_011511534.2 | Intron | XP_011509836.1 | ||||
XM_011511535.2 | Intron | XP_011509837.1 | ||||
XM_017004573.1 | Intron | XP_016860062.1 | ||||
XM_017004574.1 | Intron | XP_016860063.1 | ||||
XM_017004575.1 | Intron | XP_016860064.1 | ||||
XM_017004576.1 | Intron | XP_016860065.1 |