Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATTCTACCGTGGCCTTAAACC[C/T]TCCGAGGGGGCTTTCCCAGGCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614656 | ||||||||||||||||||||
Literature Links: |
PALD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PALD1 - phosphatase domain containing, paladin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014431.2 | Intron | NP_055246.2 | ||||
XM_011539637.2 | Intron | XP_011537939.1 | ||||
XM_011539638.2 | Intron | XP_011537940.1 | ||||
XM_017016072.1 | Intron | XP_016871561.1 |