Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTGCAGGGGTTTTGCAAAGAAA[C/T]AAGACACAGTCCCTATCATGTTAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607691 MIM: 611017 | ||||||||||||||||||||
Literature Links: |
SAR1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAR1A - secretion associated Ras related GTPase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142648.1 | Intron | NP_001136120.1 | ||||
NM_020150.4 | Intron | NP_064535.1 |
TYSND1 - trypsin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |