Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGTTGCATGGGGATTTTAAAAT[A/G]CATATTACACTTTTAATTATCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APBB1IP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APBB1IP - amyloid beta precursor protein binding family B member 1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019043.3 | Intron | NP_061916.3 | ||||
XM_006717451.2 | Intron | XP_006717514.1 | ||||
XM_011519514.2 | Intron | XP_011517816.1 |