Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGAGAAGAATCAGAAAATATCT[A/G]CCAAAAGAAGGAATAAAACAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607429 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDSS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDSS1 - prenyl (decaprenyl) diphosphate synthase, subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321978.1 | Intron | NP_001308907.1 | ||||
NM_001321979.1 | Intron | NP_001308908.1 | ||||
NM_014317.4 | Intron | NP_055132.2 | ||||
XM_011519437.2 | Intron | XP_011517739.1 | ||||
XM_017016011.1 | Intron | XP_016871500.1 |
Set Membership: |
HapMap |