Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCGTTCGTTGTTTCCTTCATTC[C/G]TTCGTCCCATGCCCTCTCGGAGCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600729 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFIC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NFIC - nuclear factor I C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001245002.1 | Intron | NP_001231931.1 | ||||
NM_001245004.1 | Intron | NP_001231933.1 | ||||
NM_001245005.1 | Intron | NP_001231934.1 | ||||
NM_005597.3 | Intron | NP_005588.2 | ||||
NM_205843.2 | Intron | NP_995315.1 | ||||
XM_005259564.3 | Intron | XP_005259621.1 | ||||
XM_006722759.2 | Intron | XP_006722822.2 | ||||
XM_017026834.1 | Intron | XP_016882323.1 | ||||
XM_017026835.1 | Intron | XP_016882324.1 | ||||
XM_017026836.1 | Intron | XP_016882325.1 |
Set Membership: |
HapMap |