Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCCAGGATGTGCCGCTGTAAGT[C/G]CCGCACCCACTCTTCCTGGATGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609475 | ||||||||||||||||||||
Literature Links: |
AKAP8L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP8L - A-kinase anchoring protein 8 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WIZ - widely interspaced zinc finger motifs | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021241.2 | 3574 | Missense Mutation | CAC,GAC | H,D 761 | NP_067064.2 | |
XM_005260004.4 | 3574 | Missense Mutation | CAC,GAC | H,D 1856 | XP_005260061.1 | |
XM_005260005.4 | 3574 | Missense Mutation | CAC,GAC | H,D 1799 | XP_005260062.1 | |
XM_005260006.4 | 3574 | Missense Mutation | CAC,GAC | H,D 1723 | XP_005260063.1 | |
XM_005260007.4 | 3574 | Missense Mutation | CAC,GAC | H,D 1666 | XP_005260064.1 | |
XM_005260008.2 | 3574 | Missense Mutation | CAC,GAC | H,D 1119 | XP_005260065.1 | |
XM_005260009.2 | 3574 | Missense Mutation | CAC,GAC | H,D 935 | XP_005260066.1 | |
XM_005260010.3 | 3574 | Missense Mutation | CAC,GAC | H,D 935 | XP_005260067.1 | |
XM_005260011.2 | 3574 | Missense Mutation | CAC,GAC | H,D 929 | XP_005260068.1 | |
XM_005260012.2 | 3574 | Missense Mutation | CAC,GAC | H,D 802 | XP_005260069.1 | |
XM_006722828.3 | 3574 | Missense Mutation | CAC,GAC | H,D 1082 | XP_006722891.1 | |
XM_011528163.2 | 3574 | Missense Mutation | CAC,GAC | H,D 935 | XP_011526465.1 |