Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTTCTCCCCCTCTACTTTTTAC[A/G]TTTCTAGTTTTATCATATTGTGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZBTB43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB43 - zinc finger and BTB domain containing 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135776.1 | Intron | NP_001129248.1 | ||||
NM_014007.3 | Intron | NP_054726.1 | ||||
XM_005251835.3 | Intron | XP_005251892.1 | ||||
XM_011518409.1 | Intron | XP_011516711.1 | ||||
XM_011518411.2 | Intron | XP_011516713.1 |