Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCGAACCCCAGCCTGCCGAGC[C/T]GCTCCTCACAGGGTGCTGTGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612027 MIM: 139139 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRASP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRASP - general receptor for phosphoinositides 1 associated scaffold protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR4A1 - nuclear receptor subfamily 4 group A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202233.1 | Intron | NP_001189162.1 | ||||
NM_001202234.1 | Intron | NP_001189163.1 | ||||
NM_002135.4 | Intron | NP_002126.2 | ||||
NM_173157.2 | Intron | NP_775180.1 | ||||
XM_005268822.3 | Intron | XP_005268879.1 | ||||
XM_005268824.3 | Intron | XP_005268881.1 | ||||
XM_006719363.1 | Intron | XP_006719426.1 | ||||
XM_006719364.3 | Intron | XP_006719427.1 | ||||
XM_017019247.1 | Intron | XP_016874736.1 | ||||
XM_017019248.1 | Intron | XP_016874737.1 | ||||
XM_017019249.1 | Intron | XP_016874738.1 |
Set Membership: |
HapMap |