Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314690 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM5C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDM5C - lysine demethylase 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146702.1 | Intron | NP_001140174.1 | ||||
NM_001282622.1 | Intron | NP_001269551.1 | ||||
NM_004187.3 | Intron | NP_004178.2 | ||||
XM_005262035.4 | Intron | XP_005262092.1 | ||||
XM_011530824.2 | Intron | XP_011529126.1 | ||||
XM_011530825.2 | Intron | XP_011529127.1 | ||||
XM_011530826.2 | Intron | XP_011529128.1 | ||||
XM_011530827.2 | Intron | XP_011529129.1 | ||||
XM_011530828.1 | Intron | XP_011529130.1 | ||||
XM_011530829.2 | Intron | XP_011529131.1 | ||||
XM_011530830.2 | Intron | XP_011529132.1 | ||||
XM_011530831.2 | Intron | XP_011529133.1 | ||||
XM_017029886.1 | Intron | XP_016885375.1 | ||||
XM_017029887.1 | Intron | XP_016885376.1 | ||||
XM_017029888.1 | Intron | XP_016885377.1 | ||||
XM_017029889.1 | Intron | XP_016885378.1 |
Set Membership: |
HapMap |