Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGGGTCCTCCCTCACCAGGAGA[G/T]ATTACTGTTTCCCCTAACACACGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610742 MIM: 609957 MIM: 165380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOV10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOV10 - Mov10 RISC complex RNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130079.2 | Intron | NP_001123551.1 | ||||
NM_001286072.1 | Intron | NP_001273001.1 | ||||
NM_001321324.1 | Intron | NP_001308253.1 | ||||
NM_020963.4 | Intron | NP_066014.1 | ||||
XM_005270869.4 | Intron | XP_005270926.1 | ||||
XM_005270870.4 | Intron | XP_005270927.1 | ||||
XM_017001318.1 | Intron | XP_016856807.1 | ||||
XM_017001319.1 | Intron | XP_016856808.1 |
PPM1J - protein phosphatase, Mg2+/Mn2+ dependent 1J | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOC - ras homolog family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042678.1 | Intron | NP_001036143.1 | ||||
NM_001042679.1 | Intron | NP_001036144.1 | ||||
NM_175744.4 | Intron | NP_786886.1 |