Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAACTCTGTATTCTGAGGCATA[A/G]ATTGGTTATAACGAATGGCAGATAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605214 | ||||||||||||||||||||
Literature Links: |
KCNMB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNMB2 - potassium calcium-activated channel subfamily M regulatory beta subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278911.1 | Intron | NP_001265840.1 | ||||
NM_005832.4 | Intron | NP_005823.1 | ||||
NM_181361.2 | Intron | NP_852006.1 | ||||
XM_011512325.2 | Intron | XP_011510627.1 |
KCNMB2-AS1 - KCNMB2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |