Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCTGCTGGAGATGGAGGGCTTA[C/G]GAGTCTCCACTGTGCAGAAAACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176390 | ||||||||||||||||||||
Literature Links: |
PSG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSG1 - pregnancy specific beta-1-glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184825.1 | 578 | Missense Mutation | CCT,CGT | P,R 147 | NP_001171754.1 | |
NM_001184826.1 | 578 | Missense Mutation | CCT,CGT | P,R 147 | NP_001171755.1 | |
NM_001297773.1 | 578 | Missense Mutation | CCT,CGT | P,R 147 | NP_001284702.1 | |
NM_006905.2 | 578 | Missense Mutation | CCT,CGT | P,R 147 | NP_008836.2 | |
XM_005259065.3 | 578 | Intron | XP_005259122.1 | |||
XM_011527125.1 | 578 | Missense Mutation | CCT,CGT | P,R 147 | XP_011525427.1 | |
XM_017026994.1 | 578 | Intron | XP_016882483.1 |