Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAACAGCCCTGGGAAATTCCAG[G/T]GATTGCTTTGTCTTTTTTTTTCTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606191 | |||||||||||||||||||||||
Literature Links: |
FNBP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
FNBP1 - formin binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR107 - G protein-coupled receptor 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136557.1 | Intron | NP_001130029.1 | ||||
NM_001136558.1 | Intron | NP_001130030.1 | ||||
NM_001287346.1 | Intron | NP_001274275.1 | ||||
NM_020960.4 | Intron | NP_066011.2 | ||||
XM_017014953.1 | Intron | XP_016870442.1 | ||||
XM_017014954.1 | Intron | XP_016870443.1 |
Set Membership: |
HapMap |