Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATACCAGGACAAAAAAACCACTA[C/T]AGACAATTAACCAGATAAAGAACTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609118 | |||||||||||||||||||||||
Literature Links: |
PDCD10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PDCD10 - programmed cell death 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007217.3 | Intron | NP_009148.2 | ||||
NM_145859.1 | Intron | NP_665858.1 | ||||
NM_145860.1 | Intron | NP_665859.1 | ||||
XM_005247086.4 | Intron | XP_005247143.1 | ||||
XM_005247087.4 | Intron | XP_005247144.1 | ||||
XM_005247088.3 | Intron | XP_005247145.1 | ||||
XM_006713485.3 | Intron | XP_006713548.1 | ||||
XM_011512368.2 | Intron | XP_011510670.1 | ||||
XM_011512369.2 | Intron | XP_011510671.1 | ||||
XM_017005644.1 | Intron | XP_016861133.1 | ||||
XM_017005645.1 | Intron | XP_016861134.1 |