Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCGCAGAGCAGTTAGTCTTTTC[A/T]TGTCTGTGAATTCAGTAATGATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604753 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF268 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF268 - zinc finger protein 268 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165881.2 | Intron | NP_001159353.1 | ||||
NM_001165882.2 | Intron | NP_001159354.1 | ||||
NM_001165883.1 | Intron | NP_001159355.1 | ||||
NM_001165884.2 | Intron | NP_001159356.2 | ||||
NM_001165885.1 | Intron | NP_001159357.1 | ||||
NM_001165886.1 | Intron | NP_001159358.1 | ||||
NM_001165887.1 | Intron | NP_001159359.1 | ||||
NM_003415.2 | Intron | NP_003406.1 | ||||
NM_152943.2 | Intron | NP_694422.2 |
Set Membership: |
HapMap |