Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATAAAAGCCCATCCATGCATCC[A/G]TGCATCCATACTTTTAAGGCAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120620 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CR1 - complement component 3b/4b receptor 1 (Knops blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000573.3 | Intron | NP_000564.2 | ||||
NM_000651.4 | Intron | NP_000642.3 | ||||
XM_006711166.2 | Intron | XP_006711229.1 | ||||
XM_011509205.1 | Intron | XP_011507507.1 |