Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGAGCACTTAATAAATCATATC[C/T]GTTGTTATTTAAAATGTTGAACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615775 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHDC7B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
KLHDC7B - kelch domain containing 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYCE3 - synaptonemal complex central element protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |