Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCATTGACCATTTTTGTGAAAA[C/T]TGACATCTTTAGGATAGTAAGTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615393 MIM: 616634 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTERF4 - mitochondrial transcription termination factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182501.3 | Intron | NP_872307.2 | ||||
XM_006712292.3 | Intron | XP_006712355.1 | ||||
XM_006712294.1 | Intron | XP_006712357.1 | ||||
XM_006712295.3 | Intron | XP_006712358.1 | ||||
XM_011510634.2 | Intron | XP_011508936.1 | ||||
XM_017003379.1 | Intron | XP_016858868.1 | ||||
XM_017003380.1 | Intron | XP_016858869.1 | ||||
XM_017003381.1 | Intron | XP_016858870.1 | ||||
XM_017003382.1 | Intron | XP_016858871.1 | ||||
XM_017003383.1 | Intron | XP_016858872.1 | ||||
XM_017003384.1 | Intron | XP_016858873.1 | ||||
XM_017003385.1 | Intron | XP_016858874.1 | ||||
XM_017003386.1 | Intron | XP_016858875.1 | ||||
XM_017003387.1 | Intron | XP_016858876.1 | ||||
XM_017003388.1 | Intron | XP_016858877.1 | ||||
XM_017003389.1 | Intron | XP_016858878.1 | ||||
XM_017003390.1 | Intron | XP_016858879.1 | ||||
XM_017003391.1 | Intron | XP_016858880.1 |
SNED1 - sushi, nidogen and EGF like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080437.1 | Intron | NP_001073906.1 | ||||
XM_011510931.2 | Intron | XP_011509233.1 | ||||
XM_011510932.2 | Intron | XP_011509234.1 | ||||
XM_011510933.2 | Intron | XP_011509235.1 | ||||
XM_011510934.2 | Intron | XP_011509236.1 | ||||
XM_011510937.2 | Intron | XP_011509239.1 | ||||
XM_017003773.1 | Intron | XP_016859262.1 | ||||
XM_017003774.1 | Intron | XP_016859263.1 | ||||
XM_017003775.1 | Intron | XP_016859264.1 | ||||
XM_017003776.1 | Intron | XP_016859265.1 | ||||
XM_017003777.1 | Intron | XP_016859266.1 | ||||
XM_017003778.1 | Intron | XP_016859267.1 | ||||
XM_017003779.1 | Intron | XP_016859268.1 | ||||
XM_017003780.1 | Intron | XP_016859269.1 | ||||
XM_017003781.1 | Intron | XP_016859270.1 | ||||
XM_017003782.1 | Intron | XP_016859271.1 |