Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGCTGTGCTGTCAGGCTTCCGGA[A/G]CAGTGCTACTCCTAGAAGCCGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616634 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNED1 - sushi, nidogen and EGF like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080437.1 | Intron | NP_001073906.1 | ||||
XM_011510931.2 | Intron | XP_011509233.1 | ||||
XM_011510932.2 | Intron | XP_011509234.1 | ||||
XM_011510933.2 | Intron | XP_011509235.1 | ||||
XM_011510934.2 | Intron | XP_011509236.1 | ||||
XM_011510937.2 | Intron | XP_011509239.1 | ||||
XM_017003773.1 | Intron | XP_016859262.1 | ||||
XM_017003774.1 | Intron | XP_016859263.1 | ||||
XM_017003775.1 | Intron | XP_016859264.1 | ||||
XM_017003776.1 | Intron | XP_016859265.1 | ||||
XM_017003777.1 | Intron | XP_016859266.1 | ||||
XM_017003778.1 | Intron | XP_016859267.1 | ||||
XM_017003779.1 | Intron | XP_016859268.1 | ||||
XM_017003780.1 | Intron | XP_016859269.1 | ||||
XM_017003781.1 | Intron | XP_016859270.1 | ||||
XM_017003782.1 | Intron | XP_016859271.1 |