Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTGATCAATCCAACAAGAAATA[A/G]GTTGAAAAATATTATTTATATCTAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 300629 | |||||||||||||||||||||||
Literature Links: |
AP1S2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AP1S2 - adaptor related protein complex 1 sigma 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272071.1 | Intron | NP_001259000.1 | ||||
NM_003916.4 | Intron | NP_003907.3 | ||||
XM_005274614.3 | Intron | XP_005274671.1 | ||||
XM_011545599.1 | Intron | XP_011543901.1 | ||||
XM_017029925.1 | Intron | XP_016885414.1 | ||||
XM_017029926.1 | Intron | XP_016885415.1 |
Set Membership: |
HapMap |