Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAGCCCCGACAGAGGCTGTGAA[C/T]GAAGTCCTAGTCAAAGACCATAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607852 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NKD2 - naked cuticle homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271082.1 | Intron | NP_001258011.1 | ||||
NM_033120.3 | Intron | NP_149111.1 | ||||
XM_005248382.3 | Intron | XP_005248439.1 | ||||
XM_005248383.3 | Intron | XP_005248440.1 | ||||
XM_011514150.2 | Intron | XP_011512452.1 | ||||
XM_017010000.1 | Intron | XP_016865489.1 |