Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCTGGAGGGCCTGCGCTCCTC[A/C]GCTAAACCCAGTTCCGGTGCTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602859 MIM: 612836 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PEX10 - peroxisomal biogenesis factor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCH2 - phospholipase C eta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303012.1 | 32 | Intron | NP_001289941.1 | |||
NM_001303013.1 | 32 | Intron | NP_001289942.1 | |||
NM_014638.3 | 32 | Intron | NP_055453.2 | |||
XM_011542455.2 | 32 | Intron | XP_011540757.1 | |||
XM_011542456.2 | 32 | UTR 5 | XP_011540758.1 | |||
XM_017002870.1 | 32 | Intron | XP_016858359.1 | |||
XM_017002871.1 | 32 | Intron | XP_016858360.1 | |||
XM_017002872.1 | 32 | Intron | XP_016858361.1 | |||
XM_017002873.1 | 32 | Intron | XP_016858362.1 | |||
XM_017002874.1 | 32 | Intron | XP_016858363.1 |
Set Membership: |
HapMap |