Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTCTGCTGCCCCGGCCTCGGC[A/C]TCCTCCGACTCCGACTCCCGGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF579 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF579 - zinc finger protein 579 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152600.2 | 647 | Missense Mutation | GAG,GAT | E,D 198 | NP_689813.2 | |
XM_017026409.1 | 647 | Missense Mutation | GAG,GAT | E,D 198 | XP_016881898.1 | |
XM_017026410.1 | 647 | Missense Mutation | GAG,GAT | E,D 198 | XP_016881899.1 | |
XM_017026411.1 | 647 | Missense Mutation | GAG,GAT | E,D 198 | XP_016881900.1 |