Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAAGAACCCTTCAGAGCCTTGC[G/A]GGTTTCTGAGCGGTACACCAGTAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603020 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFG3L1P PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AFG3L1P - AFG3 like matrix AAA peptidase subunit 1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CENPBD1 - CENPB DNA-binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145039.3 | 1645 | UTR 3 | NP_659476.2 |
DEF8 - differentially expressed in FDCP 8 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |