Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCGGAAGACCTTTGTGGCTACC[A/G]TGGTGCCGCTGGAGCAGGAGACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613198 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX6B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COX6B2 - cytochrome c oxidase subunit 6B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KMT5C - lysine methyltransferase 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032701.3 | 777 | Intron | NP_116090.2 | |||
XM_005259338.3 | 777 | Intron | XP_005259395.1 | |||
XM_006723442.3 | 777 | Intron | XP_006723505.1 | |||
XM_011527415.2 | 777 | Intron | XP_011525717.1 | |||
XM_017027388.1 | 777 | UTR 5 | XP_016882877.1 |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |