Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGGAGTTGACAGGGACACTCA[A/G]GGTGGAGCTGGGCTCCATCACTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HMCN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMCN2 - hemicentin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291815.1 | Intron | NP_001278744.1 | ||||
XM_011518465.2 | Intron | XP_011516767.1 | ||||
XM_011518466.2 | Intron | XP_011516768.1 | ||||
XM_011518467.2 | Intron | XP_011516769.1 | ||||
XM_011518468.2 | Intron | XP_011516770.1 | ||||
XM_011518469.2 | Intron | XP_011516771.1 | ||||
XM_011518470.2 | Intron | XP_011516772.1 | ||||
XM_017014585.1 | Intron | XP_016870074.1 | ||||
XM_017014586.1 | Intron | XP_016870075.1 |