Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTAATTTAGCACATGCTAAGTG[C/T]TCAGTAAATGTTAGCTAAAGTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604708 | ||||||||||||||||||||
Literature Links: |
MIR1538 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR1538 - microRNA 1538 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFAT5 - nuclear factor of activated T-cells 5, tonicity-responsive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113178.2 | Intron | NP_001106649.1 | ||||
NM_006599.3 | Intron | NP_006590.1 | ||||
NM_138713.3 | Intron | NP_619727.2 | ||||
NM_138714.3 | Intron | NP_619728.2 | ||||
NM_173214.2 | Intron | NP_775321.1 | ||||
NM_173215.2 | Intron | NP_775322.1 | ||||
XM_006721125.3 | Intron | XP_006721188.1 | ||||
XM_011522817.2 | Intron | XP_011521119.1 | ||||
XM_011522818.2 | Intron | XP_011521120.1 | ||||
XM_011522819.2 | Intron | XP_011521121.1 | ||||
XM_011522820.2 | Intron | XP_011521122.1 | ||||
XM_011522822.2 | Intron | XP_011521124.1 | ||||
XM_017022870.1 | Intron | XP_016878359.1 | ||||
XM_017022871.1 | Intron | XP_016878360.1 | ||||
XM_017022872.1 | Intron | XP_016878361.1 | ||||
XM_017022873.1 | Intron | XP_016878362.1 |