Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAAGCCTGGCCCTGGGACCAA[A/G]CGAGGAAAAAACCTCCCGCCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602359 MIM: 606684 MIM: 608303 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FXYD1 - FXYD domain containing ion transport regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278717.1 | Intron | NP_001265646.1 | ||||
NM_001278718.1 | Intron | NP_001265647.1 | ||||
NM_005031.4 | Intron | NP_005022.2 | ||||
NM_021902.3 | Intron | NP_068702.1 | ||||
XM_017026874.1 | Intron | XP_016882363.1 | ||||
XM_017026875.1 | Intron | XP_016882364.1 | ||||
XM_017026876.1 | Intron | XP_016882365.1 |
FXYD7 - FXYD domain containing ion transport regulator 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |