Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTCCCTGACCTCTGGAGCCCA[G/T]CTGAGCGGCTCCGAGAGCTCTGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609282 | ||||||||||||||||||||
Literature Links: |
MOB1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MOB1B - MOB kinase activator 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244766.1 | 102 | Intron | NP_001231695.1 | |||
NM_001244767.1 | 102 | Intron | NP_001231696.1 | |||
NM_173468.3 | 102 | Intron | NP_775739.1 | |||
XM_005265709.2 | 102 | Intron | XP_005265766.1 | |||
XM_011532412.2 | 102 | UTR 5 | XP_011530714.1 | |||
XM_017008837.1 | 102 | Intron | XP_016864326.1 |