Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTACCAGTGTGTGGTACGGCATG[A/C]GTCCTTGCATACCCCCTTGAGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613714 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCR3LG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCR3LG1 - natural killer cell cytotoxicity receptor 3 ligand 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202439.2 | 2512 | Missense Mutation | GAG,GCG | E,A 233 | NP_001189368.1 | |
XM_011520072.2 | 2512 | Missense Mutation | GAG,GCG | E,A 204 | XP_011518374.1 | |
XM_011520073.2 | 2512 | Missense Mutation | GAG,GCG | E,A 204 | XP_011518375.1 | |
XM_011520074.2 | 2512 | Missense Mutation | GAG,GCG | E,A 204 | XP_011518376.1 | |
XM_011520075.2 | 2512 | Missense Mutation | GAG,GCG | E,A 204 | XP_011518377.1 |
Set Membership: |
HapMap |