Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCACATCTTGTCCTGCTGGAAG[A/G]TCTTCAGGGGCAGTAACGGGCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615258 MIM: 604723 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL21B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
METTL21B - methyltransferase like 21B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSFM - Ts translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172695.1 | Intron | NP_001166166.1 | ||||
NM_001172696.1 | Intron | NP_001166167.1 | ||||
NM_001172697.1 | Intron | NP_001166168.1 | ||||
NM_005726.5 | Intron | NP_005717.3 |
Set Membership: |
HapMap |