Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCAGTGTTGCTAGTGAGAAGTC[C/T]GGTGCCAGTTTGAGTTTGATTCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIR5585 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR5585 - microRNA 5585 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM39B - transmembrane protein 39B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319677.1 | Intron | NP_001306606.1 | ||||
NM_001319678.1 | Intron | NP_001306607.1 | ||||
NM_001319679.1 | Intron | NP_001306608.1 | ||||
NM_018056.3 | Intron | NP_060526.2 | ||||
XM_005270988.2 | Intron | XP_005271045.1 | ||||
XM_006710724.1 | Intron | XP_006710787.1 | ||||
XM_006710725.3 | Intron | XP_006710788.1 | ||||
XM_011541681.2 | Intron | XP_011539983.1 | ||||
XM_011541683.1 | Intron | XP_011539985.1 | ||||
XM_011541684.2 | Intron | XP_011539986.1 | ||||
XM_017001595.1 | Intron | XP_016857084.1 |