Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCCGCCGCGCGCATGAGCCGGT[C/G]TTCGGGCCGTAGCGGCTCCATGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602489 | ||||||||||||||||||||
Literature Links: |
KHDRBS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KHDRBS1 - KH RNA binding domain containing, signal transduction associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271878.1 | 343 | Missense Mutation | TCT,TGT | S,C 14 | NP_001258807.1 | |
NM_006559.2 | 343 | Missense Mutation | TCT,TGT | S,C 14 | NP_006550.1 |