Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607038 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTOA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OTOA - otoancorin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161683.1 | Intron | NP_001155155.1 | ||||
NM_144672.3 | Intron | NP_653273.3 | ||||
NM_170664.2 | Intron | NP_733764.1 | ||||
XM_011545747.2 | Intron | XP_011544049.1 | ||||
XM_011545748.2 | Intron | XP_011544050.2 | ||||
XM_017022950.1 | Intron | XP_016878439.1 | ||||
XM_017022951.1 | Intron | XP_016878440.1 |
Set Membership: |
HapMap |